Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Matn2
Entrez Gene ID
17181
Gene Name
matrilin 2
Gene Aliases
Crtm2, matrilin-2
Chromosome Location
Chr.15: 34306658 - 34436242 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016762.2 NP_058042.2 14 - 15 2606 60
GenBank mRNA AK138211.1 - 14 - 15 2603 60
AK163818.1 - 14 - 15 2565 60
BC005429.1 - 14 - 15 2476 60
BC092298.1 - 14 - 15 2549 60
U69262.1 - 14 - 15 2605 60

View Full Product Details