The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_016782.2 | NP_058062.2 | 20 - 21 | 3613 | 77 |
GenBank mRNA | AF039833.1 | - | 20 - 21 | 3623 | 77 |
AK154468.1 | - | - | 4374 | 77 | |
CN538743.1 | - | 2 - 3 | 140 | 77 |