The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019428.3 | NP_062301.3 | 4 - 5 | 293 | 135 |
GenBank mRNA | AK004137.1 | - | 4 - 5 | 291 | 135 |
AK160536.1 | - | 4 - 5 | 220 | 135 | |
AK169253.1 | - | 4 - 5 | 220 | 135 | |
BC062146.1 | - | 4 - 5 | 278 | 135 | |
U95123.1 | - | 4 - 5 | 272 | 135 |