The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166531.1 | NP_001160003.1 | 15 - 16 | 1748 | 101 |
NM_001166532.1 | NP_001160004.1 | 14 - 15 | 1842 | 101 | |
NM_019460.2 | NP_062333.1 | 14 - 15 | 1699 | 101 | |
GenBank mRNA | AB032165.1 | - | 13 - 14 | 1544 | 101 |
AK154826.1 | - | 14 - 15 | 1844 | 101 | |
AK160573.1 | - | 15 - 16 | 1748 | 101 | |
BC018217.1 | - | 14 - 15 | 1710 | 101 | |
BC036972.1 | - | 14 - 15 | 1769 | 101 | |
BC066188.1 | - | 14 - 15 | 1652 | 101 |