The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001167910.1 | NP_001161382.1 | 12 - 13 | 2034 | 79 |
NM_019709.4 | NP_062683.3 | 13 - 14 | 2183 | 79 | |
GenBank mRNA | AF094820.1 | - | 11 - 12 | 1602 | 79 |
AK029048.1 | - | 12 - 13 | 2011 | 79 | |
AK122202.1 | - | 12 - 13 | 2000 | 79 | |
AK147336.1 | - | 11 - 12 | 4603 | 79 | |
AK147495.1 | - | 12 - 13 | 2008 | 79 | |
BC011533.1 | - | 12 - 13 | 1997 | 79 | |
BC054837.1 | - | 11 - 12 | 1802 | 79 | |
BC057198.1 | - | 13 - 14 | 2149 | 79 | |
DQ913734.1 | - | 11 - 12 | 1665 | 79 |