Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Snd1
Entrez Gene ID
56463
Gene Name
staphylococcal nuclease and tudor domain containing 1
Gene Aliases
AL033314, Tudor-SN
Chromosome Location
Chr.6: 28480315 - 28935162 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019776.2 NP_062750.2 12 - 13 1556 54
GenBank mRNA AB021491.1 - 12 - 13 1513 54
AK034914.1 - 11 - 12 1425 54
AK088028.1 - 12 - 13 1515 54
AK133837.1 - 12 - 13 1562 54
AK159837.1 - 12 - 13 1521 54
AK161591.1 - 12 - 13 1510 54
AK162432.1 - 12 - 13 1497 54
AK167576.1 - 12 - 13 1546 54
AK193502.1 - 2 - 3 133 54
AK201243.1 - 2 - 3 144 54
AK217790.1 - 2 - 3 135 54
BC007126.1 - 12 - 13 1503 54

View Full Product Details