The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001111331.1 | NP_001104801.1 | 3 - 4 | 694 | 93 |
NM_001291005.1 | NP_001277934.1 | 4 - 5 | 516 | 93 | |
NM_019789.4 | NP_062763.2 | 4 - 5 | 520 | 93 | |
NR_110989.1 | 4 - 5 | 680 | 93 | ||
GenBank mRNA | AF184624.1 | - | 4 - 5 | 468 | 93 |
AF274050.1 | - | 3 - 4 | 380 | 93 | |
AF287732.1 | - | 4 - 5 | 494 | 93 | |
AF287733.1 | - | 4 - 5 | 498 | 93 | |
AF300870.1 | - | 4 - 5 | 443 | 93 | |
AK016721.1 | - | 4 - 5 | 682 | 93 | |
AK165711.1 | - | 4 - 5 | 468 | 93 | |
BC026980.1 | - | 4 - 5 | 457 | 93 | |
BC047139.1 | - | 4 - 5 | 457 | 93 | |
BC057329.1 | - | 4 - 5 | 439 | 93 | |
DQ148499.1 | - | 3 - 4 | 380 | 93 | |
DQ148500.1 | - | 3 - 4 | 694 | 93 |