Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Sirt1
Entrez Gene ID
93759
Gene Name
sirtuin 1
Gene Aliases
AA673258, SIR2L1, Sir2, Sir2a, Sir2alpha
Chromosome Location
Chr.10: 63319005 - 63339061 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159589.1 NP_001153061.1 2 - 3 714 96
NM_019812.2 NP_062786.1 3 - 4 828 96
GenBank mRNA AF214646.1 - 3 - 4 813 96
AK144203.1 - 2 - 3 717 96
AY377984.1 - 3 - 4 813 96
BC152314.1 - 3 - 4 308 96

View Full Product Details