Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Mmp19
Entrez Gene ID
58223
Gene Name
matrix metallopeptidase 19
Gene Aliases
-
Chromosome Location
Chr.10: 128790910 - 128800824 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001164197.1 NP_001157669.1 7 - 8 1029 70
NM_021412.2 NP_067387.1 7 - 8 1164 70
GenBank mRNA AF153199.1 - 7 - 8 1116 70
AF155221.1 - 7 - 8 1144 70
AF162446.1 - 7 - 8 1144 70
AK079924.1 - 7 - 8 1030 70
AK080125.1 - 7 - 8 1025 70
AK086808.1 - 6 - 7 904 70
AK136517.1 - 7 - 8 1018 70
AK154863.1 - 7 - 8 1158 70
BC112966.2 - 7 - 8 1136 70

View Full Product Details