The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021491.3 | NP_067466.1 | 3 - 4 | 1793 | 78 |
GenBank mRNA | AJ250461.1 | - | 1 - 2 | 1324 | 78 |
AK019476.2 | - | 4 - 5 | 1845 | 78 | |
AK139226.1 | - | 3 - 4 | 1769 | 78 | |
BC043077.1 | - | 3 - 4 | 1793 | 78 | |
BC046980.1 | - | 2 - 3 | 1613 | 78 |