The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_021530.2 | NP_067505.2 | 24 - 25 | 3443 | 84 |
GenBank mRNA | AF224508.1 | - | 24 - 25 | 3389 | 84 |
AK034237.1 | - | 24 - 25 | 3445 | 84 | |
AK039404.1 | - | 24 - 25 | 3408 | 84 | |
AK047863.1 | - | 17 - 18 | 2377 | 84 | |
AK171610.1 | - | 23 - 24 | 3219 | 84 | |
AK173014.1 | - | 24 - 25 | 3277 | 84 | |
BC030388.1 | - | 24 - 25 | 3405 | 84 |