Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cbfb
Entrez Gene ID
12400
Gene Name
core binding factor beta
Gene Aliases
AI893578, PEA2, PEBP2b, Pebp2, Pebpb2
Chromosome Location
Chr.8: 105170674 - 105217989 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001161457.1 NP_001154929.1 2 - 3 466 84
NM_001161458.1 NP_001154930.1 2 - 3 466 84
NM_022309.4 NP_071704.3 2 - 3 466 84
GenBank mRNA BC006763.1 - 2 - 3 234 84
BC026749.1 - 2 - 3 276 84
BC040752.1 - 2 - 3 265 84
D14570.1 - 2 - 3 396 84
D14571.1 - 2 - 3 396 84
D14572.1 - 2 - 3 396 84
L03279.1 - 2 - 3 245 84
L03306.1 - - 164 84

View Full Product Details