The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146217.1 | NP_001139689.1 | 1 - 2 | 352 | 107 |
NM_022316.2 | NP_071711.2 | 1 - 2 | 352 | 107 | |
GenBank mRNA | AF070470.1 | - | 1 - 2 | 328 | 107 |
AK040931.1 | - | 1 - 2 | 353 | 107 | |
BC031804.1 | - | 1 - 2 | 332 | 107 |