The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083315.2 | NP_001076784.1 | 3 - 4 | 687 | 74 |
NM_001289624.1 | NP_001276553.1 | 3 - 4 | 611 | 74 | |
NM_001289625.1 | NP_001276554.1 | 3 - 4 | 611 | 74 | |
NM_001289626.1 | NP_001276555.1 | 3 - 4 | 611 | 74 | |
NM_001289627.1 | NP_001276556.1 | 3 - 4 | 345 | 74 | |
NM_001289629.1 | NP_001276558.1 | 3 - 4 | 345 | 74 | |
NM_022332.3 | NP_071727.2 | 3 - 4 | 687 | 74 | |
GenBank mRNA | AF234884.1 | - | 3 - 4 | 638 | 74 |
AF234885.1 | - | 3 - 4 | 638 | 74 | |
AK079107.1 | - | 3 - 4 | 688 | 74 | |
AK132103.1 | - | 3 - 4 | 641 | 74 | |
AK151550.1 | - | 3 - 4 | 642 | 74 | |
AY007792.1 | - | 3 - 4 | 573 | 74 | |
AY007793.1 | - | 3 - 4 | 573 | 74 | |
AY007794.1 | - | 4 - 5 | 602 | 74 | |
AY007795.1 | - | 4 - 5 | 602 | 74 | |
AY007796.1 | - | 4 - 5 | 412 | 74 | |
AY007797.1 | - | 4 - 5 | 412 | 74 | |
AY007798.1 | - | 3 - 4 | 345 | 74 | |
AY007799.1 | - | 3 - 4 | 345 | 74 | |
AY551821.1 | - | 3 - 4 | 338 | 74 | |
AY551822.1 | - | 3 - 4 | 338 | 74 | |
BC024652.1 | - | 3 - 4 | 611 | 74 | |
BC051429.1 | - | 3 - 4 | 465 | 74 | |
BC060630.1 | - | 3 - 4 | 528 | 74 | |
BC108418.1 | - | 3 - 4 | 631 | 74 | |
CT010245.1 | - | 3 - 4 | 264 | 74 |