Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Dpysl5
Entrez Gene ID
65254
Gene Name
dihydropyrimidinase-like 5
Gene Aliases
CRAM, CRMP-5, Crmp5
Chromosome Location
Chr.5: 30711569 - 30799369 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_023047.2 NP_075534.1 6 - 7 992 81
GenBank mRNA AF249295.1 - 6 - 7 1156 81
AK082132.1 - 3 - 4 138 81
BC065054.1 - 6 - 7 992 81

View Full Product Details