The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001136055.2 | NP_001129527.1 | 4 - 5 | 273 | 69 |
NM_001271430.1 | NP_001258359.1 | 3 - 4 | 329 | 69 | |
NM_001271431.1 | NP_001258360.1 | 3 - 4 | 197 | 69 | |
NM_001271432.1 | NP_001258361.1 | 5 - 6 | 492 | 69 | |
NM_001271461.1 | NP_001258390.1 | 4 - 5 | 284 | 69 | |
NM_001271462.1 | NP_001258391.1 | 4 - 5 | 296 | 69 | |
NM_007656.5 | NP_031682.1 | 4 - 5 | 405 | 69 | |
GenBank mRNA | AK135381.1 | - | 4 - 5 | 269 | 69 |
AK146912.1 | - | 4 - 5 | 269 | 69 | |
AK152871.1 | - | 4 - 5 | 400 | 69 | |
AK153070.1 | - | 4 - 5 | 400 | 69 | |
AK160907.1 | - | 3 - 4 | 251 | 69 | |
AK168097.1 | - | 4 - 5 | 356 | 69 | |
AK168515.1 | - | 4 - 5 | 375 | 69 | |
BC132559.1 | - | 4 - 5 | 375 | 69 | |
BC132565.1 | - | 4 - 5 | 375 | 69 | |
D14883.1 | - | 3 - 4 | 238 | 69 |