The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008052.3 | NP_032078.2 | 3 - 4 | 1813 | 98 |
GenBank mRNA | AB015422.1 | - | 2 - 3 | 1416 | 98 |
AK088630.1 | - | 3 - 4 | 1769 | 98 | |
AK169455.1 | - | 3 - 4 | 1814 | 98 | |
AK220514.1 | - | 2 - 3 | 1394 | 98 | |
BC053055.1 | - | 3 - 4 | 959 | 98 | |
BC065075.1 | - | 4 - 5 | 1595 | 98 | |
U38252.1 | - | 1 - 2 | 998 | 98 |