The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008052.3 | NP_032078.2 | 9 - 10 | 2506 | 64 |
GenBank mRNA | AB015422.1 | - | 8 - 9 | 2109 | 64 |
AK088630.1 | - | 9 - 10 | 2462 | 64 | |
AK169455.1 | - | 9 - 10 | 2507 | 64 | |
AK220514.1 | - | 8 - 9 | 2087 | 64 | |
BC053055.1 | - | 9 - 10 | 1652 | 64 | |
BC065075.1 | - | 10 - 11 | 2288 | 64 | |
U38252.1 | - | 7 - 8 | 1691 | 64 |