The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001163465.1 | NP_001156937.1 | 14 - 15 | 1914 | 106 |
NM_008164.2 | NP_032190.2 | 14 - 15 | 1860 | 106 | |
GenBank mRNA | AK085338.1 | - | - | 176 | 104 |
AK143132.1 | - | 13 - 14 | 2243 | 106 | |
AK220431.1 | - | 14 - 15 | 1831 | 106 | |
BC004843.1 | - | 10 - 11 | 1788 | 106 | |
BC052010.1 | - | 13 - 14 | 1922 | 106 | |
U43194.1 | - | 14 - 15 | 1754 | 106 |