Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Impact
Entrez Gene ID
16210
Gene Name
impact, RWD domain protein
Gene Aliases
E430016J11Rik
Chromosome Location
Chr.18: 12962943 - 12992950 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008378.2 NP_032404.1 2 - 3 252 78
GenBank mRNA AK138136.1 - 2 - 3 223 78
BC020524.1 - 2 - 3 228 78
D87973.1 - 2 - 3 252 78

View Full Product Details