Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stt3a
Entrez Gene ID
16430
Gene Name
STT3, subunit of the oligosaccharyltransferase complex, homolog A (S. cerevisiae)
Gene Aliases
AA408947, BB081708, Itm1
Chromosome Location
Chr.9: 36729345 - 36767674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008408.4 NP_032434.3 7 - 8 680 128
GenBank mRNA AK030363.1 - 7 - 8 744 128
AK077877.1 - 7 - 8 704 128
AK081547.1 - 7 - 8 736 128
AK087470.1 - 7 - 8 719 128
AK129027.1 - 7 - 8 718 128
AK150640.1 - 7 - 8 725 128
AK151541.1 - 7 - 8 736 128
AK151791.1 - 7 - 8 736 128
AK152242.1 - 7 - 8 725 128
AK153840.1 - 7 - 8 681 128
AK153886.1 - 7 - 8 781 128
AK160037.1 - 7 - 8 719 128
AK160607.1 - 7 - 8 681 128
AK168505.1 - 7 - 8 725 128
AK168937.1 - 7 - 8 725 128
AK169669.1 - 7 - 8 704 128
AK206072.1 - 3 - 4 221 128
AK207675.1 - 3 - 4 221 128
AX799090.1 - 7 - 8 723 128
BC028897.1 - 6 - 7 631 128
BC037612.1 - 7 - 8 719 128
BC085313.1 - 7 - 8 709 128
L34260.1 - 7 - 8 723 128

View Full Product Details