Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Stt3a
Entrez Gene ID
16430
Gene Name
STT3, subunit of the oligosaccharyltransferase complex, homolog A (S. cerevisiae)
Gene Aliases
AA408947, BB081708, Itm1
Chromosome Location
Chr.9: 36729345 - 36767674 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008408.4 NP_032434.3 9 - 10 1026 89
GenBank mRNA AK030363.1 - 9 - 10 1090 89
AK077877.1 - 9 - 10 1050 89
AK081547.1 - 9 - 10 1082 90
AK129027.1 - 9 - 10 1064 89
AK150640.1 - 9 - 10 1071 89
AK151541.1 - 9 - 10 1082 89
AK151791.1 - 9 - 10 1082 89
AK152242.1 - 9 - 10 1071 89
AK153840.1 - 9 - 10 1027 89
AK153886.1 - 9 - 10 1127 89
AK160037.1 - 9 - 10 1065 89
AK160607.1 - 9 - 10 1027 89
AK168505.1 - 9 - 10 1071 89
AK168937.1 - 9 - 10 1071 89
AK169669.1 - 9 - 10 1050 89
AX799090.1 - 9 - 10 1069 89
BC028897.1 - 8 - 9 977 89
BC037612.1 - 9 - 10 1065 89
BC085313.1 - 9 - 10 1055 89
L34260.1 - 9 - 10 1069 89

View Full Product Details