Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Kif4
Entrez Gene ID
16571
Gene Name
kinesin family member 4
Gene Aliases
AI323435, D330050K22Rik, Kns4
Chromosome Location
Chr.X: 100626065 - 100727271 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_008446.2 NP_032472.2 5 - 6 1087 101
GenBank mRNA AK084837.1 - 5 - 6 1064 101
AK135179.1 - 5 - 6 1035 101
AK171938.1 - 5 - 6 1000 101
BC050946.1 - 5 - 6 1025 101
BC054537.1 - 5 - 6 1009 101
D12646.1 - 5 - 6 1414 101

View Full Product Details