The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_008451.2 | NP_032477.2 | 4 - 5 | 829 | 77 |
NR_045528.1 | 4 - 5 | 866 | 77 | ||
GenBank mRNA | AF055666.1 | - | 4 - 5 | 662 | 77 |
AK141439.1 | - | 4 - 5 | 759 | 77 | |
BC014845.1 | - | 4 - 5 | 673 | 77 | |
BQ887879.1 | - | 4 - 5 | 697 | 77 |