Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dntt
Entrez Gene ID
21673
Gene Name
deoxynucleotidyltransferase, terminal
Gene Aliases
Tdt
Chromosome Location
Chr.19: 41029275 - 41059525 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_009345.2 NP_033371.2 11 - 12 1669 107
GenBank mRNA AF316014.1 - 11 - 12 1522 107
AK087978.1 - 11 - 12 1670 107
AK088709.1 - 11 - 12 1670 107
X04123.1 - 11 - 12 1553 107

View Full Product Details