The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166584.1 | NP_001160056.1 | 1 - 2 | 404 | 66 |
NM_009346.3 | NP_033372.1 | 1 - 2 | 404 | 66 | |
GenBank mRNA | AK140307.1 | - | 1 - 2 | 405 | 66 |
AK148375.1 | - | 1 - 2 | 370 | 66 | |
L13853.1 | - | 1 - 2 | 131 | 66 | |
S74227.1 | - | 1 - 2 | 404 | 66 |