The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009517.2 | NP_033543.2 | 1 - 2 | 161 | 99 |
GenBank mRNA | AF012923.1 | - | 1 - 2 | 115 | 99 |
BQ895699.1 | - | 1 - 2 | 174 | 99 |