The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009536.4 | NP_033562.3 | 4 - 5 | 716 | 96 |
GenBank mRNA | AF483478.1 | - | 4 - 5 | 575 | 96 |
AF483479.1 | - | 4 - 5 | 575 | 96 | |
AK011612.1 | - | 1 - 2 | 85 | 96 | |
AK075995.1 | - | 4 - 5 | 665 | 96 | |
AK135831.1 | - | 4 - 5 | 644 | 96 | |
AK135867.1 | - | 4 - 5 | 644 | 96 | |
AK169812.1 | - | 4 - 5 | 671 | 96 | |
AK188179.1 | - | 2 - 3 | 341 | 96 | |
AK190793.1 | - | 2 - 3 | 328 | 96 | |
AK191008.1 | - | 2 - 3 | 306 | 96 | |
AK192790.1 | - | 2 - 3 | 320 | 96 | |
AK195513.1 | - | 2 - 3 | 320 | 96 | |
AK201131.1 | - | 1 - 2 | 96 | ||
AK218242.1 | - | 2 - 3 | 294 | 96 | |
BC058686.1 | - | 4 - 5 | 675 | 96 | |
D87663.1 | - | 4 - 5 | 603 | 96 | |
Z19599.1 | - | 4 - 5 | 658 | 96 |