The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009538.2 | NP_033564.2 | 5 - 6 | 709 | 68 |
GenBank mRNA | AF147785.1 | - | 2 - 3 | 402 | 68 |
AF324471.1 | - | 1 - 2 | 311 | 68 | |
AK142052.1 | - | 6 - 7 | 701 | 68 | |
AK142210.1 | - | 7 - 8 | 738 | 68 | |
BC065150.1 | - | 5 - 6 | 709 | 68 | |
BC141284.1 | - | 6 - 7 | 551 | 68 | |
BC145612.1 | - | 6 - 7 | 551 | 68 | |
FJ425893.1 | - | - | 1026 | 68 | |
X95503.1 | - | 7 - 8 | 688 | 68 | |
X95504.1 | - | 1 - 2 | 242 | 68 |