The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010352.2 | NP_034482.2 | 2 - 3 | 261 | 103 |
GenBank mRNA | CA465575.1 | - | - | 251 | 103 |
D87325.1 | - | 2 - 3 | 140 | 103 |