Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Invs
Entrez Gene ID
16348
Gene Name
inversin
Gene Aliases
Nphp2, inv
Chromosome Location
Chr.4: 48279760 - 48431953 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001281977.1 NP_001268906.1 1 - 2 218 86
NM_001281978.1 NP_001268907.1 1 - 2 218 86
NM_010569.4 NP_034699.3 1 - 2 218 86
GenBank mRNA AF034860.3 - 1 - 2 160 86
AJ010902.1 - 1 - 2 159 86
AK020257.1 - 1 - 2 171 86
AK029087.1 - 1 - 2 154 86
AK029305.1 - 1 - 2 163 86
AK040307.1 - 1 - 2 219 86
AK041671.1 - 1 - 2 153 86
AK158766.1 - 1 - 2 177 86
BC126929.1 - 1 - 2 107 86
BX528321.1 - 1 - 2 199 86

View Full Product Details