Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Krtap19-5
Entrez Gene ID
16704
Gene Name
keratin associated protein 19-5
Gene Aliases
Krtap8-2
Chromosome Location
Chr.16: 88895968 - 88896449 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010676.2 NP_034806.2 1 - 1 63 85
GenBank mRNA AA717139.1 - 1 - 1 71 85
D86422.1 - 1 - 1 74 85

View Full Product Details