Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lamp1
Entrez Gene ID
16783
Gene Name
lysosomal-associated membrane protein 1
Gene Aliases
AI196048, CD107a, Lamp-1
Chromosome Location
Chr.8: 13159134 - 13175338 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_010684.2 NP_034814.2 8 - 9 1339 84
GenBank mRNA AK004637.1 - 8 - 9 1206 84
AK159096.1 - 8 - 9 1205 84
AK159376.1 - 8 - 9 1178 84
AK172007.1 - 8 - 9 1221 84
AY069968.1 - 8 - 9 1276 84
BC006044.1 - 6 - 7 881 84
BC006785.1 - 4 - 5 436 84
BC049097.1 - 8 - 9 1313 84
J03881.1 - 7 - 8 1013 84
M25244.1 - 8 - 9 1194 84
M32015.1 - 8 - 9 1163 84

View Full Product Details