The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_010739.2 | NP_034869.1 | 3 - 4 | 886 | 118 |
GenBank mRNA | AK132274.1 | - | 2 - 3 | 731 | 118 |
BC024321.1 | - | 2 - 3 | 852 | 118 | |
J04634.1 | - | 3 - 4 | 886 | 118 |