Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tle1
Entrez Gene ID
21885
Gene Name
transducin-like enhancer of split 1, homolog of Drosophila E(spl)
Gene Aliases
C230057C06Rik, Estm14, Grg-1, Grg1, Tle4l
Chromosome Location
Chr.4: 72109945 - 72200893 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001285529.1 NP_001272458.1 9 - 10 1595 111
NM_001285530.1 NP_001272459.1 7 - 8 1376 111
NM_011599.5 NP_035729.3 9 - 10 1598 111
GenBank mRNA AK048515.1 - 9 - 10 1585 111
AK052961.1 - 7 - 8 1699 111
AK082499.1 - 9 - 10 1564 111
AY155197.1 - 7 - 8 534 111
AY155198.1 - 5 - 6 397 111
AY155199.1 - 9 - 10 757 111
AY155200.1 - 3 - 4 231 111
BC057573.1 - 9 - 10 1425 111
U61362.1 - 9 - 10 764 111

View Full Product Details