The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011766.5 | NP_035896.1 | 1 - 2 | 547 | 63 |
GenBank mRNA | AF107306.1 | - | 1 - 2 | 68 | 63 |
AF118845.1 | - | 1 - 2 | 317 | 63 | |
AF125166.1 | - | 1 - 2 | 548 | 63 | |
AK033131.1 | - | 1 - 2 | 239 | 63 | |
AK046521.1 | - | 1 - 2 | 555 | 63 | |
BC059241.1 | - | 1 - 2 | 447 | 63 |