Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Zfpm2
Entrez Gene ID
22762
Gene Name
zinc finger protein, multitype 2
Gene Aliases
B330005D23Rik, FOG-2, FOG2
Chromosome Location
Chr.15: 40655042 - 41104592 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011766.5 NP_035896.1 7 - 8 1473 90
GenBank mRNA AF107306.1 - 7 - 8 994 90
AF118845.1 - 7 - 8 1243 90
AF125166.1 - 7 - 8 1474 90
BC059241.1 - 7 - 8 1373 90
EF514219.1 - 7 - 8 2296 90

View Full Product Details