The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_011834.2 | NP_035964.1 | 7 - 8 | 889 | 67 |
GenBank mRNA | AF072376.1 | - | 7 - 8 | 889 | 67 |
AK075578.1 | - | 7 - 8 | 67 | ||
AK144042.1 | - | 7 - 8 | 842 | 67 | |
BC012637.1 | - | 7 - 8 | 892 | 67 |