Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Vnn3
Entrez Gene ID
26464
Gene Name
vanin 3
Gene Aliases
-
Chromosome Location
Chr.10: 23851462 - 23869843 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_011979.2 NP_036109.3 2 - 3 462 71
GenBank mRNA AJ132103.1 - 2 - 3 452 71
BC111521.2 - 2 - 3 434 71

View Full Product Details