Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Tinag
Entrez Gene ID
26944
Gene Name
tubulointerstitial nephritis antigen
Gene Aliases
AI452335, TIN-ag
Chromosome Location
Chr.9: 76951687 - 77045806 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_012033.3 NP_036163.3 3 - 4 584 123
GenBank mRNA AF153366.1 - 3 - 4 526 123
BC010745.1 - 3 - 4 529 123

View Full Product Details