The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001083319.1 | NP_001076788.1 | 10 - 11 | 1442 | 82 |
NM_013699.2 | NP_038727.1 | 9 - 10 | 1334 | 82 | |
GenBank mRNA | AB221581.1 | - | 8 - 9 | 922 | 82 |
AK079815.1 | - | 2 - 3 | 169 | 82 | |
AK140942.1 | - | 4 - 5 | 339 | 82 | |
AY182062.1 | - | 9 - 10 | 1031 | 82 | |
BC034749.1 | - | 8 - 9 | 1661 | 82 | |
BC139828.1 | - | 8 - 9 | 951 | 82 | |
BC141082.1 | - | 8 - 9 | 1013 | 82 | |
CD806234.1 | - | 4 - 5 | 408 | 82 | |
CF172242.1 | - | 4 - 5 | 436 | 82 | |
U20086.1 | - | 8 - 9 | 925 | 82 |