Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Abcc2
Entrez Gene ID
12780
Gene Name
ATP-binding cassette, sub-family C (CFTR/MRP), member 2
Gene Aliases
AI173996, Abc30, Cmoat, Mrp2, cMRP
Chromosome Location
Chr.19: 43782308 - 43838332 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_013806.2 NP_038834.2 4 - 5 561 85
GenBank mRNA AF227274.1 - 4 - 5 465 85
AF282772.1 - 4 - 5 465 85
AF282773.1 - 4 - 5 465 85

View Full Product Details