Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Incenp
Entrez Gene ID
16319
Gene Name
inner centromere protein
Gene Aliases
2700067E22Rik, AU019509, C130081E20, C77457
Chromosome Location
Chr.19: 9872297 - 9899546 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_016692.3 NP_057901.2 5 - 6 1248 100
GenBank mRNA AB100432.1 - 4 - 5 1042 100
AB100433.1 - 4 - 5 1042 100
AF117610.1 - 5 - 6 1184 100
AK034764.1 - 5 - 6 1177 100
AK081841.1 - 5 - 6 1182 100
AK088627.1 - 5 - 6 1205 100
BC021761.2 - 5 - 6 1145 100
BC037011.1 - 5 - 6 1201 100
BC052414.1 - 5 - 6 1181 100

View Full Product Details