The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_018765.3 | NP_061235.2 | 4 - 5 | 603 | 90 |
GenBank mRNA | AF071184.1 | - | 4 - 5 | 455 | 90 |
AK135215.1 | - | 4 - 5 | 603 | 90 | |
AK159522.1 | - | 4 - 5 | 855 | 90 | |
AK160559.1 | - | 4 - 5 | 340 | 90 | |
AK161496.1 | - | 4 - 5 | 297 | 90 | |
AK162009.1 | - | 4 - 5 | 345 | 90 | |
BC034851.1 | - | 4 - 5 | 298 | 90 |