Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Lgals12
Entrez Gene ID
56072
Gene Name
lectin, galactose binding, soluble 12
Gene Aliases
AW987437, Grip1
Chromosome Location
Chr.19: 7596656 - 7610466 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019516.3 NP_062389.1 1 - 2 520 78
GenBank mRNA AF223223.1 - 2 - 3 214 78
AF244978.1 - 2 - 3 222 78
AF244979.1 - 1 - 2 267 78
AK033535.1 - 1 - 2 447 78
AK136526.1 - 1 - 2 521 78
BC030890.1 - 1 - 2 204 78

View Full Product Details