The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019516.3 | NP_062389.1 | 6 - 7 | 1011 | 72 |
GenBank mRNA | AF223223.1 | - | 7 - 8 | 705 | 72 |
AF244978.1 | - | 7 - 8 | 713 | 72 | |
AF244979.1 | - | 6 - 7 | 758 | 72 | |
AK085660.1 | - | 6 - 7 | 1014 | 72 | |
AK136526.1 | - | 6 - 7 | 1012 | 72 | |
BC030890.1 | - | 6 - 7 | 695 | 72 |