The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_019686.5 | NP_062660.1 | 3 - 4 | 261 | 90 |
GenBank mRNA | AB016080.1 | - | 3 - 4 | 261 | 90 |
AK005347.1 | - | 3 - 4 | 365 | 90 | |
AK013240.1 | - | - | 370 | 90 | |
BC005739.1 | - | 3 - 4 | 303 | 90 |