The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252401.1 | NP_001239330.1 | 11 - 12 | 1231 | 134 |
NM_019725.2 | NP_062699.1 | 11 - 12 | 1333 | 134 | |
GenBank mRNA | AF145958.2 | - | 11 - 12 | 1088 | 134 |
AK220533.1 | - | 10 - 11 | 2142 | 134 | |
BC051955.1 | - | 10 - 11 | 1286 | 134 | |
BC062961.1 | - | 10 - 11 | 1286 | 134 | |
BC071219.1 | - | 11 - 12 | 1235 | 134 |