The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001291161.1 | NP_001278090.1 | 3 - 4 | 486 | 67 |
NM_019818.5 | NP_062792.2 | 3 - 4 | 567 | 67 | |
NM_023355.2 | NP_075844.1 | 2 - 3 | 370 | 67 | |
NR_111896.1 | 3 - 4 | 475 | 67 | ||
GenBank mRNA | AF223950.1 | - | 3 - 4 | 533 | 67 |
AK011745.1 | - | 3 - 4 | 519 | 67 | |
AK012130.2 | - | 3 - 4 | 521 | 67 | |
AK017389.1 | - | 3 - 4 | 520 | 67 | |
AK028132.1 | - | 3 - 4 | 519 | 67 | |
AK153837.1 | - | 3 - 4 | 522 | 67 | |
BC022610.1 | - | 3 - 4 | 503 | 67 | |
BC055279.1 | - | 3 - 4 | 531 | 67 | |
CA751902.1 | - | 3 - 4 | 459 | 67 | |
CF740208.1 | - | 2 - 3 | 330 | 67 |