Assay Design

The assay primers and probes lie within a single exon. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Dnajb8
Entrez Gene ID
56691
Gene Name
DnaJ (Hsp40) homolog, subfamily B, member 8
Gene Aliases
1700016F14Rik, 1700031F10Rik, AI326313, mDj6
Chromosome Location
Chr.6: 88222268 - 88223256 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_019964.1 NP_064348.1 1 - 1 47 77
GenBank mRNA AB028856.1 - 1 - 1 47 77

View Full Product Details